1-(4-Bromo-3-chlorobenzyl)piperidine ≥95%

Supplier: AOBChem USA

Warning

24960-500MG 24960-1G 24960-5G 24960-25G
77324-616EA 213.21 USD
77324-616 77348-462 77360-946 77384-300
1-(4-Bromo-3-chlorobenzyl)piperidine ≥95%
1-(4-Bromo-3-chlorobenzyl)piperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrClN
MW: 288.611 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22628249
CAS Number: 1414870-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR