1-(2,6-Dibromobenzyl)pyrrolidine ≥95%

Supplier: AOBChem USA

Warning

24959-500MG 24959-1G 24959-5G 24959-25G
77324-614EA 222.48 USD
77324-614 77348-460 77360-944 77384-298
1-(2,6-Dibromobenzyl)pyrrolidine ≥95%
1-(2,6-Dibromobenzyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₁H₁₃Br₂N
Storage Temperature: Ambient
MDL Number: MFCD22628248
CAS Number: 1414870-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR