4-(2,6-Dibromobenzyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

24955-500MG 24955-1G 24955-5G 24955-25G
77324-612EA 222.48 USD
77324-612 77348-458 77360-942 77384-296
4-(2,6-Dibromobenzyl)morpholine ≥97%
4-(2,6-Dibromobenzyl)morpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₃Br₂NO
Storage Temperature: Ambient
MDL Number: MFCD22628245
CAS Number: 1414870-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR