4-(3-Bromo-4-methylbenzyl)morpholine ≥95%

Supplier: AOBChem USA

Warning

24950-500MG 24950-1G 24950-5G 24950-25G
77324-608EA 222.48 USD
77324-608 77348-454 77360-938 77384-292
4-(3-Bromo-4-methylbenzyl)morpholine ≥95%
4-(3-Bromo-4-methylbenzyl)morpholine

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD22628240
CAS Number: 1414870-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR