4-Bromo-2-(1H-tetrazol-5-yl)-phenylamine ≥97%

Supplier: AOBChem USA

Warning

24944-500MG 24944-1G 24944-5G 24944-25G
77324-606EA 151.41 USD
77324-606 77345-416 77360-936 77384-290
4-Bromo-2-(1H-tetrazol-5-yl)-phenylamine ≥97%
4-Bromo-2-(1H-tetrazol-5-yl)-phenylamine

Precaución: for Research Use Only

Formula: C₇H₆BrN₅
MW: 240.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17215772
CAS Number: 27398-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR