2-Fluoro-3-iodo-5-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

24904-500MG 24904-1G 24904-5G 24904-25G
77324-602EA 266.77 USD
77324-602 77348-450 77360-934 77384-286
2-Fluoro-3-iodo-5-phenylpyridine ≥97%
2-Fluoro-3-iodo-5-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₇FIN
MW: 299.083 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14701844
CAS Number: 928822-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR