2-Fluoro-5-(4-(trifluoromethoxy)phenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

24903-500MG 24903-1G 24903-5G 24903-25G
77324-600EA 266.77 USD
77324-600 77348-448 77360-932 77384-284
2-Fluoro-5-(4-(trifluoromethoxy)phenyl)pyridine ≥97%
2-Fluoro-5-(4-(trifluoromethoxy)phenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₇F₄NO
MW: 257.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18406387
CAS Number: 1261592-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR