5-Cyclopropyl-2-fluoro-3-hydroxypyridine ≥95%

Supplier: AOBChem USA

Warning

24897-500MG 24897-1G 24897-5G
77339-970EA 695.25 USD
77339-970 77348-444 77372-716
5-Cyclopropyl-2-fluoro-3-hydroxypyridine ≥95%
5-Cyclopropyl-2-fluoro-3-hydroxypyridine

Precaución: for Research Use Only

Formula: C₈H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD28977162
CAS Number: 2624417-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR