5-Cyclopropyl-2-fluoro-3-iodopyridine ≥95%

Supplier: AOBChem USA

Warning

24893-500MG 24893-1G 24893-5G 24893-25G
77324-596EA 247.2 USD
77324-596 77348-442 77360-928 77384-280
5-Cyclopropyl-2-fluoro-3-iodopyridine ≥95%
5-Cyclopropyl-2-fluoro-3-iodopyridine

Precaución: for Research Use Only

Formula: C₈H₇FIN
MW: 263.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28154564
CAS Number: 1034467-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR