5-Cylclopropyl-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

24886-500MG 24886-1G 24886-5G 24886-25G
77324-594EA 176.13 USD
77324-594 77346-508 77360-926 77384-278
5-Cylclopropyl-2-methoxypyridine ≥97%
5-Cylclopropyl-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₉H₁₁NO
MW: 149.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22628236
CAS Number: 1063960-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR