4-Chloro-3-isopropylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

24866-500MG 24866-1G 24866-25G 24866-5G
77324-592EA 289.43 USD
77324-592 77348-440 77384-276 77372-714
4-Chloro-3-isopropylbenzoic acid ≥95%
4-Chloro-3-isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
MW: 198.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19686195
CAS Number: 1349716-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR