5-Fluoro-4-formyl-3-methoxyphenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

24845-500MG 24845-1G 24845-5G
77337-806EA 445.99 USD
77337-806 77348-436 77372-710
5-Fluoro-4-formyl-3-methoxyphenylboronic acid pinacol ester ≥95%
5-Fluoro-4-formyl-3-methoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BFO₄
MW: 280.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619417
CAS Number: 2555544-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR