1-[[2-bromo-5-(trifluoromethyl)phenyl]methyl]-piperidine ≥97%
Supplier: AOBChem USA
|
Warning
|
24836-500MG
24836-1G
24836-5G
24836-25G
77324-586EA
177.16
USD
77324-586
77346-578
77360-918
77384-270
1-[[2-bromo-5-(trifluoromethyl)phenyl]methyl]-piperidine ≥97%
1-[[2-bromo-5-(trifluoromethyl)phenyl]methyl]-piperidine
Precaución: for Research Use Only
Formula:
C₁₃H₁₅BrF₃N MW: 322.164 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22628223 CAS Number: 913296-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...