2,6-Difluorothioanisole ≥97%

Supplier: AOBChem USA

Warning

24758-500MG 24758-1G 24758-5G 24758-25G
77324-582EA 141.11 USD
77324-582 77344-780 77360-914 77384-266
2,6-Difluorothioanisole ≥97%
2,6-Difluorothioanisole

Precaución: for Research Use Only

Formula: C₇H₆F₂S
MW: 160.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185576
CAS Number: 91524-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR