Methyl 3,5-difluoro-4-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

24755-500MG 24755-1G 24755-5G 24755-25G
77324-580EA 258.53 USD
77324-580 77348-430 77360-912 77384-264
Methyl 3,5-difluoro-4-(methylthio)benzoate ≥97%
Methyl 3,5-difluoro-4-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD34755806
CAS Number: 2115373-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR