4-[(2-Bromo-5-methylphenyl)methyl]-morpholine ≥97%

Supplier: AOBChem USA

Warning

24740-500MG 24740-1G 24740-5G 24740-25G
77324-578EA 278.1 USD
77324-578 77348-428 77360-910 77384-262
4-[(2-Bromo-5-methylphenyl)methyl]-morpholine ≥97%
4-[(2-Bromo-5-methylphenyl)methyl]-morpholine

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO
MW: 270.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22628212
CAS Number: 1414870-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR