4-((5-Bromopyridin-3-yl)methyl)morpholine ≥95%

Supplier: AOBChem USA

Warning

24736-500MG 24736-1G 24736-5G 24736-25G
77324-576EA 244.11 USD
77324-576 77348-426 77360-908 77384-260
4-((5-Bromopyridin-3-yl)methyl)morpholine ≥95%
4-((5-Bromopyridin-3-yl)methyl)morpholine

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrN₂O
MW: 257.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18260426
CAS Number: 364793-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR