Methyl 3-ethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

24720-500MG 24720-1G 24720-5G 24720-25G
77324-574EA 170.98 USD
77324-574 77346-392 77360-906 77384-258
Methyl 3-ethylbenzoate ≥97%
Methyl 3-ethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04037896
CAS Number: 50604-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR