1-[(4-Bromo-2-chlorophenyl)carbonyl]pyrrolidine ≥97%

Supplier: AOBChem USA

Warning

24714-1G 24714-5G 24714-25G
77342-110EA 114.33 USD
77342-110 77360-904 77372-708
1-[(4-Bromo-2-chlorophenyl)carbonyl]pyrrolidine ≥97%
1-[(4-Bromo-2-chlorophenyl)carbonyl]pyrrolidine

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrClNO
MW: 288.568 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20046462
CAS Number: 877383-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR