2-(6-(Methoxymethoxy)naphthalen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%

Supplier: AOBChem USA

Warning

24702-500MG 24702-1G 24702-5G 24702-25G
77324-570EA 280.16 USD
77324-570 77348-422 77360-902 77384-256
2-(6-(Methoxymethoxy)naphthalen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%
2-(6-(Methoxymethoxy)naphthalen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₈H₂₃BO₄
Storage Temperature: Ambient
MDL Number: MFCD22493687
CAS Number: 874673-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR