2-(Trimethylsilyl)benzofuran ≥97%

Supplier: AOBChem USA

Warning

24627-500MG 24627-1G 24627-5G 24627-25G
77324-566EA 120.51 USD
77324-566 77343-666 77360-898 77372-704
2-(Trimethylsilyl)benzofuran ≥97%
2-(Trimethylsilyl)benzofuran

Precaución: for Research Use Only

Formula: C₁₁H₁₄OSi
MW: 190.314 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28659943
CAS Number: 60981-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR