(6-Fluoro-5-methylpyridin-3-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

24592-500MG 24592-1G 24592-25G 24592-5G
77324-562EA 289.43 USD
77324-562 77348-416 77384-252 77372-700
(6-Fluoro-5-methylpyridin-3-yl)methanol ≥95%
(6-Fluoro-5-methylpyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₈FNO
MW: 141.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520710
CAS Number: 1260776-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR