4-Methyl-6-methylsulfanyl-pyridine-3-carbaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
24583-500MG
24583-1G
24583-5G
77324-560EA
327.54
USD
77324-560
77348-414
77372-698
4-Methyl-6-methylsulfanyl-pyridine-3-carbaldehyde ≥97%
4-Methyl-6-methylsulfanyl-pyridine-3-carbaldehyde
Precaución: for Research Use Only
Formula:
C₈H₉NOS MW: 167.228 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21088014 CAS Number: 1289056-48-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...