1-Bromo-3-fluoro-2-methoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

24576-1G 24576-5G 24576-25G
77341-404EA 85.49 USD
77341-404 77360-894 77372-696
1-Bromo-3-fluoro-2-methoxy-5-methylbenzene ≥97%
1-Bromo-3-fluoro-2-methoxy-5-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO
MW: 219.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543733
CAS Number: 1394291-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR