2-Methoxy-4-(methylsulfanyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

24574-500MG 24574-1G 24574-5G 24574-25G
77324-556EA 198.79 USD
77324-556 77348-410 77360-892 77384-248
2-Methoxy-4-(methylsulfanyl)pyridine ≥95%
2-Methoxy-4-(methylsulfanyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₉NOS
Storage Temperature: Ambient
MDL Number: MFCD17011772
CAS Number: 1378609-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR