2-Chloro-4-(1-piperidinylcarbonyl)phenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
24543-500MG
24543-1G
24543-25G
24543-5G
77324-554EA
289.43
USD
77324-554
77348-406
77384-246
77372-692
2-Chloro-4-(1-piperidinylcarbonyl)phenylboronic acid ≥97%
2-Chloro-4-(1-piperidinylcarbonyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₁₂H₁₅BClNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD22125178 CAS Number: 1451392-96-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...