4-Methyl-2-(methylsulfanyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

24522-500MG 24522-1G 24522-5G
77324-552EA 321.36 USD
77324-552 77348-404 77372-688
4-Methyl-2-(methylsulfanyl)benzoic acid ≥97%
4-Methyl-2-(methylsulfanyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₂S
Storage Temperature: Ambient
MDL Number: MFCD16853043
CAS Number: 24852-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR