2-Chloro-4-Bromo-5-fluorobenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-Chloro-4-bromo-5-fluorobenzaldehyde, 4-Bromo-2-chloro-5-fluorobenzaldehyde
24518-1G
24518-5G
24518-25G
77340-608EA
60.77
USD
77340-608
77360-890
77372-686
2-Chloro-4-Bromo-5-fluorobenzaldehyde ≥97%
2-Chloro-4-Bromo-5-fluorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₃BrClFO MW: 237.452 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13185877 CAS Number: 1214386-29-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...