4-(4-Bromo-3-methoxybenzyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

24468-500MG 24468-1G 24468-5G 24468-25G
77324-548EA 201.88 USD
77324-548 77348-400 77360-886 77384-242
4-(4-Bromo-3-methoxybenzyl)morpholine ≥97%
4-(4-Bromo-3-methoxybenzyl)morpholine

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO₂
MW: 286.165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543706
CAS Number: 1394291-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR