4-[[2-Bromo-5-(trifluoromethyl)phenyl]methyl]morpholine ≥97%

Supplier: AOBChem USA

Warning

24467-500MG 24467-1G 24467-5G 24467-25G
77324-546EA 181.28 USD
77324-546 77346-778 77360-884 77384-240
4-[[2-Bromo-5-(trifluoromethyl)phenyl]methyl]morpholine ≥97%
4-[[2-Bromo-5-(trifluoromethyl)phenyl]methyl]morpholine

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrF₃NO
MW: 324.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543705
CAS Number: 1394291-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR