1-Benzyl-5-bromopyridin-2(1H)-one ≥97%

Supplier: AOBChem USA

Warning

24466-1G 24466-5G 24466-25G
77340-886EA 62.83 USD
77340-886 77360-882 77372-684
1-Benzyl-5-bromopyridin-2(1H)-one ≥97%
1-Benzyl-5-bromopyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO
Storage Temperature: Ambient
MDL Number: MFCD02326300
CAS Number: 217448-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR