2,4-Dibromo-6-chloro-1-isobutoxybenzene ≥97%

Supplier: AOBChem USA

Warning

24423-500MG 24423-1G 24423-5G 24423-25G
77324-544EA 222.48 USD
77324-544 77348-398 77360-876 77384-238
2,4-Dibromo-6-chloro-1-isobutoxybenzene ≥97%
2,4-Dibromo-6-chloro-1-isobutoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁Br₂ClO
MW: 342.455 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543704
CAS Number: 1394291-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR