1,5-Dibromo-3-chloro-2-(phenylmethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

24422-500MG 24422-1G 24422-5G 24422-25G
77324-542EA 222.48 USD
77324-542 77348-396 77360-874 77384-236
1,5-Dibromo-3-chloro-2-(phenylmethoxy)benzene ≥97%
1,5-Dibromo-3-chloro-2-(phenylmethoxy)benzene

Precaución: for Research Use Only

Formula: C₁₃H₉Br₂ClO
MW: 376.471 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543703
CAS Number: 864181-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR