2,4-Dibromo-6-chloro-1-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

24421-500MG 24421-1G 24421-5G 24421-25G
77324-540EA 222.48 USD
77324-540 77348-394 77360-872 77384-234
2,4-Dibromo-6-chloro-1-isopropoxybenzene ≥97%
2,4-Dibromo-6-chloro-1-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉Br₂ClO
Storage Temperature: Ambient
MDL Number: MFCD22543702
CAS Number: 1394291-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR