1-[(2-Bromo-5-methoxyphenyl)methyl]piperidine ≥95%

Supplier: AOBChem USA

Warning

24375-500MG 24375-1G 24375-5G 24375-25G
77324-536EA 199.82 USD
77324-536 77348-392 77360-864 77384-230
1-[(2-Bromo-5-methoxyphenyl)methyl]piperidine ≥95%
1-[(2-Bromo-5-methoxyphenyl)methyl]piperidine

Precaución: for Research Use Only

Formula: C₁₃H₁₈BrNO
MW: 284.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20994298
CAS Number: 1394291-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR