Methyl 4-chloro-3-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

24371-500MG 24371-1G 24371-5G 24371-25G
77324-534EA 150.38 USD
77324-534 77345-334 77360-862 77384-228
Methyl 4-chloro-3-isopropoxybenzoate ≥97%
Methyl 4-chloro-3-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₃
MW: 228.672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19440114
CAS Number: 1394933-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR