4-Chloro-3-(propan-2-yloxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24369-500MG 24369-1G 24369-5G 24369-25G
77324-532EA 187.46 USD
77324-532 77348-390 77360-860 77384-226
4-Chloro-3-(propan-2-yloxy)benzaldehyde ≥97%
4-Chloro-3-(propan-2-yloxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
MW: 198.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19440116
CAS Number: 1289135-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR