(4-Chloro-3-ethoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

24366-500MG 24366-1G 24366-5G 24366-25G
77324-530EA 204.97 USD
77324-530 77348-388 77360-858 77384-224
(4-Chloro-3-ethoxyphenyl)methanol ≥97%
(4-Chloro-3-ethoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD15527076
CAS Number: 909853-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR