1,2-Dibromo-3,4,5-trifluorobenzene ≥97%

Supplier: AOBChem USA

Warning

24338-500MG 24338-1G 24338-5G 24338-25G
77324-528EA 90.64 USD
77324-528 77342-378 77360-856 77372-672
1,2-Dibromo-3,4,5-trifluorobenzene ≥97%
1,2-Dibromo-3,4,5-trifluorobenzene

Precaución: for Research Use Only

Formula: C₆HBr₂F₃
MW: 289.875 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070782
CAS Number: 17299-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR