5-Bromo-N-methoxypyridine-3-carboxamide ≥97%

Supplier: AOBChem USA

Warning

24074-500MG 24074-1G 24074-5G 24074-25G
77324-518EA 266.77 USD
77324-518 77348-376 77360-844 77384-214
5-Bromo-N-methoxypyridine-3-carboxamide ≥97%
5-Bromo-N-methoxypyridine-3-carboxamide

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
MW: 231.047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17831441
CAS Number: 1137063-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR