3-Methyl-2,5-bis(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

24008-500MG 24008-1G 24008-5G
77337-392EA 414.06 USD
77337-392 77348-374 77372-658
3-Methyl-2,5-bis(methylthio)pyridine ≥97%
3-Methyl-2,5-bis(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₈H₁₁NS₂
MW: 185.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755795
CAS Number: 1227384-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR