3-Methyl-2-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

24004-500MG 24004-1G 24004-5G 24004-25G
77324-514EA 121.54 USD
77324-514 77343-904 77360-838 77372-656
3-Methyl-2-(methylthio)pyridine ≥97%
3-Methyl-2-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₇H₉NS
MW: 139.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22666567
CAS Number: 51933-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR