5-Iodo-3-methyl-2-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

24003-500MG 24003-1G 24003-25G 24003-5G
77324-512EA 289.43 USD
77324-512 77348-372 77384-210 77372-654
5-Iodo-3-methyl-2-(methylthio)pyridine ≥97%
5-Iodo-3-methyl-2-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₇H₈INS
MW: 265.115 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27756650
CAS Number: 1809158-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR