5-Methyl-6-methylsulfanyl-nicotinic acid ≥97%

Supplier: AOBChem USA

Warning

24002-500MG 24002-1G 24002-5G 24002-25G
77324-510EA 226.6 USD
77324-510 77348-370 77360-836 77384-208
5-Methyl-6-methylsulfanyl-nicotinic acid ≥97%
5-Methyl-6-methylsulfanyl-nicotinic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₂S
Storage Temperature: Ambient
MDL Number: MFCD21087240
CAS Number: 1355226-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR