3-Bromo-4-phenoxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

23999-500MG 23999-1G 23999-5G 23999-25G
77324-508EA 199.82 USD
77324-508 77348-368 77360-834 77384-206
3-Bromo-4-phenoxybenzonitrile ≥97%
3-Bromo-4-phenoxybenzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₈BrNO
MW: 274.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18906613
CAS Number: 183111-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR