3-Bromo-4-(4-chlorophenoxy)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

23995-500MG 23995-1G 23995-5G 23995-25G
77324-506EA 242.05 USD
77324-506 77348-366 77360-832 77384-204
3-Bromo-4-(4-chlorophenoxy)benzonitrile ≥97%
3-Bromo-4-(4-chlorophenoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₇BrClNO
MW: 308.558 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25801500
CAS Number: 1355656-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR