5-Methyl-6-methylsulfanyl-pyridine-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

23962-500MG 23962-1G 23962-5G 23962-25G
77324-504EA 245.14 USD
77324-504 77348-364 77360-830 77384-202
5-Methyl-6-methylsulfanyl-pyridine-3-carbaldehyde ≥97%
5-Methyl-6-methylsulfanyl-pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₉NOS
MW: 167.228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21087235
CAS Number: 1289077-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR