2,3-Difluoro-6-(methylthio)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

23954-500MG 23954-1G 23954-5G 23954-25G
77324-500EA 245.14 USD
77324-500 77348-362 77360-826 77384-198
2,3-Difluoro-6-(methylthio)benzaldehyde ≥95%
2,3-Difluoro-6-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂OS
Storage Temperature: Ambient
MDL Number: MFCD22543679
CAS Number: 1394291-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR