6-Fluoro-2-(methylthio)benzoic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

23948-500MG 23948-1G 23948-5G 23948-25G
77324-498EA 208.06 USD
77324-498 77348-360 77360-822 77384-196
6-Fluoro-2-(methylthio)benzoic acid methyl ester ≥97%
6-Fluoro-2-(methylthio)benzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
Storage Temperature: Ambient
MDL Number: MFCD30289725
CAS Number: 1865064-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR