2-Bromo-5-(piperidin-1-ylmethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

23920-500MG 23920-1G 23920-25G 23920-5G
77324-496EA 289.43 USD
77324-496 77348-358 77384-194 77372-650
2-Bromo-5-(piperidin-1-ylmethyl)pyridine ≥97%
2-Bromo-5-(piperidin-1-ylmethyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrN₂
MW: 255.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18073299
CAS Number: 364794-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR